Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243925.1 | NP_001230854.1 | 3 | 423 | ||
NM_001243926.1 | NP_001230855.1 | 5 | 902 | |||
NM_004635.4 | NP_004626.1 | 3 | 408 | |||
GenBank mRNA | AB451303.1 | - | 2 | 260 | ||
AK290969.1 | - | 3 | 379 | |||
AK313565.1 | - | 2 | 325 | |||
BC001662.2 | - | 3 | 369 | 2988489 | ||
BC007591.1 | - | 3 | 345 | 3345889 | ||
BC010407.2 | - | 3 | 367 | 3833667 | ||
BC068497.1 | - | 5 | 856 | |||
BM848086.1 | - | 3 | 423 | |||
U09578.1 | - | 3 | 351 | |||
U43784.1 | - | 2 | 282 |