Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058209.2 | NP_478116.2 | 234 | |||
XM_008763803.1 | XP_008762025.1 | 3 | 531 | |||
XM_008763804.1 | XP_008762026.1 | 3 | 528 | |||
GenBank mRNA | AF072439.1 | - | 259 | |||
BC166844.1 | - | 171 |