Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078617.3 | NP_511172.1 | 3 | 259 | ||
GenBank mRNA | BC058134.1 | - | 3 | 263 | 6918484 | |
FQ221811.1 | - | 3 | 251 | |||
FQ222488.1 | - | 3 | 249 | |||
FQ222494.1 | - | 3 | 253 | |||
FQ222496.1 | - | 3 | 250 | |||
FQ222641.1 | - | 3 | 251 | |||
FQ223677.1 | - | 3 | 251 | |||
FQ228659.1 | - | 3 | 251 | |||
FQ228931.1 | - | 3 | 251 |