Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080580.2 | NP_542147.1 | 4 | 537 | ||
XM_006242593.1 | XP_006242655.1 | 4 | 533 | |||
XM_006242594.2 | XP_006242656.1 | 4 | 653 | |||
GenBank mRNA | BC081741.1 | - | 4 | 537 | 7110109 | |
M83681.1 | - | 3 | 365 | |||
U90206.1 | - | 3 | 428 |