Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080688.1 | NP_542419.1 | 9 | 1389 | ||
XM_006245135.2 | XP_006245197.1 | 10 | 2464 | |||
XM_006245136.2 | XP_006245198.1 | 9 | 1305 | |||
XM_006245137.2 | XP_006245199.1 | 9 | 1604 | |||
XM_006245138.2 | XP_006245200.1 | 9 | 1479 | |||
XM_008767182.1 | XP_008765404.1 | 9 | 1883 | |||
XM_008767183.1 | XP_008765405.1 | 9 | 1883 | |||
XM_008767184.1 | XP_008765406.1 | 9 | 1922 | |||
XM_008767185.1 | XP_008765407.1 | 9 | 1306 | |||
XM_008767186.1 | XP_008765408.1 | 9 | 1884 | |||
XM_008767187.1 | XP_008765409.1 | 9 | 1884 | |||
XM_008767188.1 | XP_008765410.1 | 9 | 1883 | |||
XM_008767189.1 | XP_008765411.1 | 9 | 1883 | |||
XM_008767190.1 | XP_008765412.1 | 9 | 1882 | |||
XM_008767191.1 | XP_008765413.1 | 9 | 1882 | |||
GenBank mRNA | U16655.1 | - | 9 | 1389 |