Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080787.1 | NP_542965.1 | 18 | 1948 | ||
XM_008765012.1 | XP_008763234.1 | 17 | 2950 | |||
XM_008765013.1 | XP_008763235.1 | 17 | 2950 | |||
XM_008765014.1 | XP_008763236.1 | 17 | 2860 | |||
XM_008765015.1 | XP_008763237.1 | 17 | 2792 | |||
XM_008765016.1 | XP_008763238.1 | 17 | 2033 | |||
XM_008765017.1 | XP_008763239.1 | 18 | 2818 | |||
XM_008765018.1 | XP_008763240.1 | 18 | 2728 | |||
XM_008765019.1 | XP_008763241.1 | 18 | 2727 | |||
XM_008765020.1 | XP_008763242.1 | 18 | 2654 | |||
GenBank mRNA | FQ209812.1 | - | 18 | 1842 | ||
S49760.1 | - | 18 | 1948 |