Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080885.1 | NP_543161.1 | 6 | 438 | ||
GenBank mRNA | L02121.1 | - | 6 | 438 |