Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005387.6 | NP_005378.4 | 3 | 577 | ||
NM_016320.4 | NP_057404.2 | 3 | 577 | |||
NM_139131.4 | NP_624357.1 | 3 | 577 | |||
NM_139132.3 | NP_624358.2 | 3 | 577 | |||
GenBank mRNA | AB040538.1 | - | 3 | 332 | ||
AF071076.1 | - | 3 | 280 | |||
AF071077.1 | - | 3 | 280 | |||
AF231130.1 | - | 3 | 300 | |||
AK292882.1 | - | 3 | 317 | |||
AK301512.1 | - | 3 | 337 | |||
BC041136.1 | - | 3 | 326 | 5179730 | ||
BG773331.1 | - | 3 | 501 | |||
KU847959.1 | - | 156 | ||||
U41815.1 | - | 3 | 300 |