Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp26a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130408.2 | NP_569092.2 | 2 | 269 | ||
GenBank mRNA | AF439720.2 | - | 2 | 269 | ||
DQ317305.1 | - | 2 | 279 |