Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LATS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004690.3 | NP_004681.1 | 8 | 4011 | ||
NR_073033.1 | 9 | 3917 | ||||
XM_017011474.1 | XP_016866963.1 | 9 | 3952 | |||
XM_017011475.1 | XP_016866964.1 | 9 | 3872 | |||
XM_017011476.1 | XP_016866965.1 | 10 | 4020 | |||
XM_017011478.1 | XP_016866967.1 | 9 | 3930 | |||
XM_017011479.1 | XP_016866968.1 | 9 | 3977 | |||
XM_017011480.1 | XP_016866969.1 | 8 | 3649 | |||
XM_017011481.1 | XP_016866970.1 | 7 | 3695 | |||
GenBank mRNA | AF104413.1 | - | 8 | 4011 | ||
AF164041.1 | - | 7 | 3495 |