Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130751.1 | NP_570107.1 | 1, 2 | 191 | ||
GenBank mRNA | AB078902.1 | - | 1, 2 | 191 |