Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130756.3 | NP_570112.3 | 1, 2 | 530 | ||
GenBank mRNA | AF452100.1 | - | 1, 2 | 180 | ||
BC078751.1 | - | 1, 2 | 524 | 7114090 | ||
CV115391.1 | - | 1, 2 | 530 |