Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080898.2 | NP_543174.1 | 1 | 170 | ||
GenBank mRNA | AY064411.1 | - | 501 | |||
EF672039.1 | - | 1 | 170 |