Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130402.2 | NP_569086.1 | 2 | 186 | ||
XM_008763309.1 | XP_008761531.1 | 3 | 683 | |||
GenBank mRNA | AF321552.1 | - | 4 | 372 | ||
EF100688.1 | - | 2 | 186 |