Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130416.2 | NP_569100.2 | 7 | 761 | ||
XM_006251624.2 | XP_006251686.1 | 8 | 788 | |||
XM_006251625.2 | XP_006251687.1 | 8 | 2587 | |||
XM_006251626.2 | XP_006251688.1 | 9 | 2668 | |||
XM_006251627.2 | XP_006251689.1 | 7 | 720 | |||
XM_008770484.1 | XP_008768706.1 | 8 | 969 | |||
XM_008770485.1 | XP_008768707.1 | 7 | 902 | |||
GenBank mRNA | AF280423.1 | - | 753 | |||
BC070896.1 | - | 7 | 761 | 7099644 |