Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130404.1 | NP_569088.1 | 3 | 284 | ||
XM_006249629.1 | XP_006249691.1 | 4 | 364 | |||
XM_008769420.1 | XP_008767642.1 | 4 | 340 | |||
GenBank mRNA | AF105329.1 | - | 3 | 284 | ||
FQ228843.1 | - | 4 | 346 |