Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130406.1 | NP_569090.1 | 7, 8 | 911 | ||
GenBank mRNA | AF293459.1 | - | 7, 8 | 911 |