Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318807.1 | NP_001305736.1 | 8 | 2196 | ||
NM_014430.2 | NP_055245.2 | 7 | 2047 | |||
XM_011536659.2 | XP_011534961.1 | 7 | 1703 | |||
XM_017021221.1 | XP_016876710.1 | 7 | 3751 | |||
GenBank mRNA | AF190901.1 | - | 7 | 2091 | ||
AF218586.1 | - | 5 | 1225 | |||
AF544398.1 | - | 5 | 1008 | |||
AK000387.1 | - | 5 | 1001 | |||
AK122828.1 | - | 7 | 3639 | |||
BC035970.1 | - | 5 | 1035 | 4716269 |