Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130416.2 | NP_569100.2 | 10, 11 | 1256 | ||
XM_006251624.2 | XP_006251686.1 | 11, 12 | 1283 | |||
XM_006251625.2 | XP_006251687.1 | 11, 12 | 3082 | |||
XM_006251626.2 | XP_006251688.1 | 12, 13 | 3163 | |||
XM_006251627.2 | XP_006251689.1 | 10, 11 | 1215 | |||
XM_008770484.1 | XP_008768706.1 | 11, 12 | 1464 | |||
XM_008770485.1 | XP_008768707.1 | 10, 11 | 1397 | |||
GenBank mRNA | AF280423.1 | - | 1248 | |||
BC070896.1 | - | 10, 11 | 1256 | 7099644 |