Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130735.1 | NP_570091.1 | 6, 7 | 588 | ||
XM_008762547.1 | XP_008760769.1 | 6, 7 | 1534 | |||
GenBank mRNA | Y08981.1 | - | 6, 7 | 588 |