Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128160.1 | NP_001121632.1 | 2, 3 | 803 | ||
NM_001128161.1 | NP_001121633.1 | 3, 4 | 422 | |||
NM_014517.4 | NP_055332.3 | 2, 3 | 803 | |||
GenBank mRNA | AF198487.1 | - | 2, 3 | 447 | ||
AK293100.1 | - | 3, 4 | 422 | |||
BC047235.1 | - | 2, 3 | 802 | 5262788 | ||
DB131031.1 | - | 3, 4 | 395 |