Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cysltr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133413.1 | NP_596904.1 | 1 | 73 | ||
XM_008770842.1 | XP_008769064.1 | 3 | 781 | |||
XM_008770843.1 | XP_008769065.1 | 4 | 2211 | |||
XM_008770844.1 | XP_008769066.1 | 3 | 2140 | |||
GenBank mRNA | AB052661.1 | - | 1 | 73 |
Set Membership: |
G-protein coupled receptor |