Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133414.1 | NP_596905.1 | 2, 3 | 188 | ||
XM_006233127.1 | XP_006233189.1 | 2, 3 | 227 | |||
XM_006233128.2 | XP_006233190.1 | 2, 3 | 286 | |||
XM_008761365.1 | XP_008759587.1 | 2, 3 | 286 | |||
GenBank mRNA | AB052921.1 | - | 1, 2 | 146 | ||
AB052922.1 | - | 2, 3 | 188 | |||
AB052923.1 | - | 2, 3 | 208 | |||
AF117330.1 | - | 2, 3 | 176 | |||
BC081736.1 | - | 2, 3 | 264 | 7128078 |