Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014555.3 | NP_055370.1 | 3, 4 | 482 | ||
XM_017017628.1 | XP_016873117.1 | 4, 5 | 569 | |||
GenBank mRNA | AF177473.1 | - | 3, 4 | 482 | ||
BC093787.1 | - | 3, 4 | 479 | 7939632 | ||
BC093789.1 | - | 3, 4 | 479 | 7939634 | ||
BC143351.1 | - | 3, 4 | 479 | 9051856 | ||
BC143353.1 | - | 3, 4 | 479 | 9051858 |