Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133518.1 | NP_598202.1 | 5 | 412 | ||
XM_006249379.2 | XP_006249441.1 | 5 | 344 | |||
XM_006249380.2 | XP_006249442.1 | 5 | 649 | |||
XM_006249381.2 | XP_006249443.1 | 5 | 648 | |||
GenBank mRNA | U12571.1 | - | 5 | 412 |