Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133324.1 | NP_579858.1 | 4, 5 | 344 | ||
XM_006253526.2 | XP_006253588.1 | 4, 5 | 609 | |||
XM_006253527.1 | XP_006253589.1 | 2, 3 | 208 | |||
GenBank mRNA | AF272892.1 | - | 4, 5 | 344 | ||
BC074005.1 | - | 4, 5 | 321 | 7099269 |