Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103689943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245686.2 | XP_006245748.1 | 3 | 2038 | ||
XM_006245687.2 | XP_006245749.1 | 3 | 964 | |||
XM_006245688.1 | XP_006245750.1 | 3 | 816 | |||
XM_008767447.1 | XP_008765669.1 | 3 | 964 | |||
XM_008767448.1 | XP_008765670.1 | 2 | 1471 |
Gene Symbol: Marf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133421.2 | NP_596912.1 | 3 | 990 | ||
XM_003750779.3 | XP_003750827.1 | 3 | 2037 | |||
XM_006245854.2 | XP_006245916.1 | 3 | 964 | |||
XM_006245855.2 | XP_006245917.1 | 3 | 816 | |||
XM_008767442.1 | XP_008765664.1 | 2 | 1471 | |||
GenBank mRNA | AB012133.1 | - | 3 | 990 |