Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000431.3 | NP_000422.1 | 3 | 398 | ||
NM_001114185.2 | NP_001107657.1 | 3 | 389 | |||
NM_001301182.1 | NP_001288111.1 | 3 | 398 | |||
XM_011538372.2 | XP_011536674.2 | 3 | 506 | |||
XM_017019313.1 | XP_016874802.1 | 3 | 351 | |||
XM_017019314.1 | XP_016874803.1 | 3 | 864 | |||
GenBank mRNA | AB209722.1 | - | 3 | 281 | ||
AF217536.1 | - | 2 | 194 | |||
AK293130.1 | - | 3 | 266 | |||
AK295338.1 | - | 3 | 256 | |||
AK315678.1 | - | 3 | 277 | |||
BC016140.1 | - | 3 | 241 | 3921543 | ||
DC421549.1 | - | 3 | 398 | |||
KU178205.1 | - | 2 | 205 | |||
KU178206.1 | - | 2 | 205 | |||
M88468.1 | - | 3 | 296 |