Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133304.1 | NP_579838.1 | 13 | 2216 | ||
XM_006257046.2 | XP_006257108.1 | 14 | 2516 | |||
XM_006257048.2 | XP_006257110.1 | 14 | 2751 | |||
XM_006257049.2 | XP_006257111.1 | 13 | 2551 | |||
XM_008773248.1 | XP_008771470.1 | 15 | 3149 | |||
XM_008773249.1 | XP_008771471.1 | 15 | 2845 | |||
XM_008773250.1 | XP_008771472.1 | 14 | 2519 | |||
GenBank mRNA | AF246120.1 | - | 13 | 2216 |