Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc40a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133315.2 | NP_579849.2 | 6 | 835 | ||
GenBank mRNA | AF394785.3 | - | 6 | 780 | ||
CK364414.1 | - | 5 | 467 | |||
FQ226522.1 | - | 5 | 495 | |||
U76714.1 | - | 6 | 850 |
Gene Symbol: LOC100911874 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750695.3 | XP_003750743.1 | 1258 |