Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270807.1 | NP_001257736.1 | 3 | 633 | ||
NM_001270808.1 | NP_001257737.1 | 3 | 682 | |||
NM_001270809.1 | NP_001257738.1 | 3 | 583 | |||
NM_001270810.1 | NP_001257739.1 | 3 | 1269 | |||
NM_001270811.1 | NP_001257740.1 | 3 | 1619 | |||
NM_133386.3 | NP_596877.2 | 2 | 1873 | |||
XM_006247787.2 | XP_006247849.1 | 3 | 953 | |||
XM_006247788.2 | XP_006247850.1 | 3 | 464 | |||
GenBank mRNA | AB049571.1 | - | 3 | 681 | ||
AB049572.1 | - | 3 | 633 | |||
AB049573.1 | - | 3 | 580 | |||
AB049574.1 | - | 3 | 1239 | |||
AB049575.1 | - | 2 | 1386 | |||
BC081738.1 | - | 2 | 1838 | 7132896 | ||
DQ486894.1 | - | 3 | 421 |