Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133393.2 | NP_596884.1 | 2, 3 | 545 | ||
GenBank mRNA | AB054539.1 | - | 2, 3 | 486 | ||
BC070933.1 | - | 2, 3 | 545 | 7097855 |