Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133398.1 | NP_596889.1 | 4, 5 | 1065 | ||
XM_006241520.2 | XP_006241582.1 | 4, 5 | 2484 | |||
XM_006241524.2 | XP_006241586.1 | 4, 5 | 2482 | |||
GenBank mRNA | AF100421.2 | - | 4, 5 | 1065 | ||
FQ226094.1 | - | 4, 5 | 1061 | |||
FQ227915.1 | - | 2, 3 | 213 |