Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133428.2 | NP_596919.1 | 7 | 939 | ||
GenBank mRNA | AB055895.1 | - | 7 | 939 | ||
AB055896.1 | - | 7 | 930 | |||
AF194029.1 | - | 7 | 894 | |||
BC089779.1 | - | 7 | 928 | 7371893 | ||
FQ219046.1 | - | 5 | 543 |