Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133526.1 | NP_598210.1 | 5 | 657 | ||
GenBank mRNA | BC061785.1 | - | 5 | 513 | 5623788 | |
FQ214867.1 | - | 5 | 565 | |||
FQ215876.1 | - | 1 | 29 | |||
FQ216712.1 | - | 5 | 686 | |||
FQ220073.1 | - | 1 | 29 | |||
FQ220093.1 | - | 1 | 29 | |||
FQ227464.1 | - | 5 | 674 | |||
FQ229225.1 | - | 1 | 29 | |||
FQ229522.1 | - | 5 | 575 | |||
Y13275.2 | - | 5 | 657 |