Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133543.3 | NP_598227.3 | 522 | |||
GenBank mRNA | BC088090.1 | - | 522 | 7368214 | ||
BC098650.1 | - | 584 | 7371059 | |||
FQ209576.1 | - | 506 | ||||
FQ209710.1 | - | 540 | ||||
U18762.1 | - | 731 | ||||
U33501.1 | - | 503 |