Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198850.1 | NP_001185779.1 | 3 | 481 | ||
NM_001198851.1 | NP_001185780.1 | 5 | 524 | |||
NM_001198852.1 | NP_001185781.1 | 3 | 386 | |||
NM_014947.4 | NP_055762.3 | 3 | 403 | |||
XM_005270632.4 | XP_005270689.1 | 2 | 291 | |||
XM_006710458.3 | XP_006710521.1 | 3 | 510 | |||
XM_006710459.3 | XP_006710522.1 | 2 | 292 | |||
XM_011541026.2 | XP_011539328.1 | 4 | 455 | |||
XM_017000693.1 | XP_016856182.1 | 3 | 530 | |||
GenBank mRNA | AB028964.1 | - | 5 | 524 | ||
AK027075.1 | - | 3 | 481 | |||
BC014182.2 | - | 3 | 386 | |||
BC151828.1 | - | 3 | 372 | 40148791 | ||
BC152441.1 | - | 5 | 524 | 8862565 |