Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyatl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134330.2 | NP_599157.2 | 2, 3 | 129 | ||
GenBank mRNA | AB019693.1 | - | 195 | |||
BC078701.1 | - | 2, 3 | 129 | 7130696 |
Gene Symbol: Glyatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126278.1 | NP_001119750.1 | 1, 2 | 90 | ||
GenBank mRNA | BC160862.1 | - | 1, 2 | 90 | 7323546 | |
FQ210014.1 | - | 244 | ||||
FQ219712.1 | - | 1, 2 | 93 |