Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133568.1 | NP_598252.1 | 3 | 1369 | ||
GenBank mRNA | AF134409.1 | - | 3 | 1369 |
Gene Symbol: Tmem87a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762124.1 | XP_008760346.1 | 20 | 1859 | ||
XM_008762125.1 | XP_008760347.1 | 20 | 1856 |