Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133586.2 | NP_598270.1 | 185 | |||
XM_008760588.1 | XP_008758810.1 | 3 | 154 | |||
XM_008760589.1 | XP_008758811.1 | 2 | 160 | |||
GenBank mRNA | AB010635.1 | - | 185 | |||
AB191005.1 | - | 222 |
Gene Symbol: LOC100910040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749155.3 | XP_003749203.1 | 2 | 250 |