Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133597.1 | NP_598281.1 | 3, 4 | 452 | ||
GenBank mRNA | AY574259.1 | - | 3, 4 | 275 | ||
J04636.1 | - | 3, 4 | 452 |