Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133608.2 | NP_598292.2 | 2 | 480 | ||
GenBank mRNA | AF239262.1 | - | 1 | 314 | ||
BC097285.1 | - | 2 | 480 | 7441748 | ||
BC097286.1 | - | 2 | 422 | 7444297 |