Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133623.1 | NP_598307.1 | 2, 3 | 335 | ||
XM_006237218.2 | XP_006237280.1 | 2, 3 | 319 | |||
XM_006237219.2 | XP_006237281.1 | 3, 4 | 435 | |||
XM_006237220.2 | XP_006237282.1 | 2, 3 | 319 | |||
XM_006237221.2 | XP_006237283.1 | 3, 4 | 435 | |||
XM_006237223.2 | XP_006237285.1 | 1, 2 | 318 | |||
GenBank mRNA | M95762.1 | - | 2, 3 | 335 |