Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBD1 SIRNA Assays ›
Gene Symbol:
MBD1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
CXXC3, PCM1, RFT
Chromosome Location:
Chr.18: 50266882 - 50281774 on Build GRCh38
UniGene ID:
Hs.405610
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5729
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204136.1
NP_001191065.1
3
644
NM_001204137.1
NP_001191066.1
3
644
NM_001204138.1
NP_001191067.1
3
644
NM_001204139.1
NP_001191068.1
3
644
NM_001204140.1
NP_001191069.1
3
644
NM_001204141.1
NP_001191070.1
3
644
NM_001204142.1
NP_001191071.1
3
644
NM_001204143.1
NP_001191072.1
3
425
NM_001204151.2
NP_001191080.1
3
644
NM_001323942.1
NP_001310871.1
3
644
NM_001323947.1
NP_001310876.1
3
644
NM_001323949.1
NP_001310878.1
3
636
NM_001323950.1
NP_001310879.1
3
644
NM_001323951.1
NP_001310880.1
3
644
NM_001323952.1
NP_001310881.1
3
644
NM_001323953.1
NP_001310882.1
3
651
NM_001323954.1
NP_001310883.1
3
644
NM_002384.2
NP_002375.1
3
644
NM_015844.2
NP_056669.2
3
644
NM_015845.3
NP_056670.2
3
644
NM_015846.3
NP_056671.2
3
644
NM_015847.3
NP_056723.2
3
644
XM_005258271.2
XP_005258328.1
3
391
XM_006722456.2
XP_006722519.1
3
391
XM_011525991.1
XP_011524293.1
3
391
XM_011525993.2
XP_011524295.1
3
391
XM_011525994.2
XP_011524296.1
3
391
XM_011525998.2
XP_011524300.1
3
391
XM_011525999.1
XP_011524301.1
3
391
XM_011526001.1
XP_011524303.1
3
391
XM_011526002.1
XP_011524304.1
3
391
XM_011526003.2
XP_011524305.1
3
391
XM_011526006.1
XP_011524308.1
3
391
XM_011526007.1
XP_011524309.1
3
391
XM_017025751.1
XP_016881240.1
3
391
XM_017025752.1
XP_016881241.1
3
391
XM_017025753.1
XP_016881242.1
3
391
XM_017025754.1
XP_016881243.1
3
391
XM_017025755.1
XP_016881244.1
3
391
XM_017025756.1
XP_016881245.1
3
391
XM_017025757.1
XP_016881246.1
3
391
XM_017025758.1
XP_016881247.1
3
391
XM_017025759.1
XP_016881248.1
3
391
XM_017025760.1
XP_016881249.1
3
391
XM_017025761.1
XP_016881250.1
3
391
XM_017025762.1
XP_016881251.1
3
391
XM_017025763.1
XP_016881252.1
3
391
XM_017025764.1
XP_016881253.1
3
391
XM_017025765.1
XP_016881254.1
3
391
XM_017025766.1
XP_016881255.1
3
391
XM_017025767.1
XP_016881256.1
3
391
XM_017025768.1
XP_016881257.1
3
391
XM_017025769.1
XP_016881258.1
3
391
XM_017025770.1
XP_016881259.1
3
391
XM_017025771.1
XP_016881260.1
3
391
XM_017025772.1
XP_016881261.1
3
391
XM_017025773.1
XP_016881262.1
3
391
XM_017025774.1
XP_016881263.1
3
391
XM_017025775.1
XP_016881264.1
3
391
XM_017025776.1
XP_016881265.1
3
391
XM_017025777.1
XP_016881266.1
3
391
GenBank mRNA
AF078830.1
-
2
207
AF078831.1
-
2
207
AF078832.1
-
2
207
AF078833.1
-
2
207
AJ564845.1
-
3
346
AK225889.1
-
3
554
AK225948.1
-
3
350
AK291519.1
-
3
645
AK295401.1
-
3
376
AK300759.1
-
3
425
AK302004.1
-
3
336
BC033242.1
-
3
370
5432943
BC047353.1
-
3
329
EF488685.1
-
2
207
Y10746.1
-
3
346
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
methylation-dependent chromatin silencing
transcription from RNA polymerase II promoter
heart development
aging
response to fluoxetine
neuron differentiation
response to nutrient levels
response to estradiol
response to cocaine
regulation of DNA methylation
negative regulation of transcription, DNA-templated
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
methyl-CpG binding
double-stranded methylated DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0