Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133538.2 | NP_598222.2 | 8 | 888 | ||
XM_006257427.2 | XP_006257489.1 | 7 | 756 | |||
XM_006257428.2 | XP_006257490.1 | 7 | 874 | |||
XM_006257430.2 | XP_006257492.1 | 6 | 687 | |||
XM_006257431.2 | XP_006257493.1 | 4 | 395 | |||
XM_008773388.1 | XP_008771610.1 | 7 | 1028 | |||
XM_008773389.1 | XP_008771611.1 | 8 | 915 | |||
XM_008773390.1 | XP_008771612.1 | 4 | 489 | |||
XM_008773391.1 | XP_008771613.1 | 4 | 1096 | |||
GenBank mRNA | AF448818.1 | - | 8 | 888 |