Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133545.1 | NP_598229.1 | 8 | 958 | ||
GenBank mRNA | U17971.1 | - | 8 | 958 |