Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751248.3 | XP_003751296.1 | 759 |
Gene Symbol: Pla2g4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133551.2 | NP_598235.2 | 17 | 2159 | ||
GenBank mRNA | BC070940.1 | - | 17 | 2159 | 7099813 | |
U38376.1 | - | 17 | 2160 |