Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133553.2 | NP_598237.2 | 1 | 1209 | ||
GenBank mRNA | AB003478.1 | - | 1 | 1180 | ||
BC126083.1 | - | 1 | 1209 | 8375818 |