Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133603.2 | NP_598287.1 | 2 | 522 | ||
XM_006248023.2 | XP_006248085.1 | 2 | 948 | |||
XM_008768554.1 | XP_008766776.1 | 2 | 758 | |||
GenBank mRNA | AF071003.1 | - | 2 | 363 | ||
BC089778.1 | - | 2 | 522 | 7301610 |