Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133617.2 | NP_598301.2 | 3, 4 | 1089 | ||
GenBank mRNA | BC088113.1 | - | 3, 4 | 1089 | 7326099 | |
U55765.1 | - | 2, 3 | 1001 |
Gene Symbol: LOC100909524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750227.3 | XP_003750275.1 | 3, 4 | 1107 |